Phosphate Buffer Mixed pH 4,0

Supplier: ACTIVE MOTIF

BP556
ACSDBP556EA 129 EUR
ACSDBP556
Phosphate Buffer Mixed pH 4,0
Buffers pH Calibration Buffers

Shelf Life on Ship Date: 12 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.


Certifications: ISO 9001.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR