Strontium standard solution, 1000 mg/L Sr in dil. nitric acid standard for ICP
Supplier: ACTIVE MOTIF
C056.2NP.L1 C056.2NP.L25 C056.2NP.L5
ACSDC056.2NP.L1EA 93.4 EUR
ACSDC056.2NP.L1 ACSDC056.2NP.L25 ACSDC056.2NP.L5
Strontium standard solution, 1000 mg/L Sr in dil. nitric acid standard for ICP
Standards Cationic Standards Strontium Standards