TK competent cells
Supplier: AGILENT
200134
AGLS200134EA 634 EUR
AGLS200134
TK competent cells
Competent Cells
Easily induce and express phosphorylated proteins, which can be used to screen expression libraries, to affinity purify or blot proteins that interact with the tyrosine-phosphorylated target.
  • Competent cell lines for inducible phosphoprotein expression
TKB1 Competent Cells are derived from the BL21 strain and are intended for use with pET, pCAL or other T7 RNA promoter-based vectors. TKX1 Competent Cells are derived from the XL1-Blue strain and are intended for use with non-T7 promoter-based vectors.
The BL21(DE3) TK strain, abbreviated TKB1,More Product Information