Standard hydrometers
Supplier: ALLA ATELIERS
3100MC08020BEO 3100MC15020HEO 3500LC10020MEU 3500LC08520BEU 3100MC08020BEU 3100MC09020BEU 3100MC10020MEU 3100MC13020HEU 3100MC14020HEU 3100MC16020HEU 3100MC11020MEU 3100MC12020MEU 3100MC13020MEU 3500MC07020BEU 3500MC07520BEU 3500MC08020BEU 3500MC08520BEU 3500MC09020BEU 3500MC14520BEU 3500MC06520BEU 3500MC14520HEU
ALLA3100MC08020BEOEA 1010 EUR
ALLA3100MC08020BEO ALLA3100MC15020HEO ALLA3500LC10020MEU ALLA3500LC08520BEU ALLA3100MC08020BEU ALLA3100MC09020BEU ALLA3100MC10020MEU ALLA3100MC13020HEU ALLA3100MC14020HEU ALLA3100MC16020HEU ALLA3100MC11020MEU ALLA3100MC12020MEU ALLA3100MC13020MEU ALLA3500MC07020BEU ALLA3500MC07520BEU ALLA3500MC08020BEU ALLA3500MC08520BEU ALLA3500MC09020BEU ALLA3500MC14520BEU ALLA3500MC06520BEU ALLA3500MC14520HEU
Standard hydrometers
Hydrometers
These standard hydrometers strictly adhere to the principles of manufacture and specified calibration.
  • Reading below meniscus
  • Calibration temperature of 20 °C
  • Reference mark on the stem shows any movement of the measurement scale
Certifications: NF, ISO, BS and DIN.
Delivery information: Supplied with certificate of conformity to ISO 649 (corresponding to BS718, DIN12791, NF B 35511).