Glass fiber filters without binder resin
Supplier: Merck
AP4001000 AP4002405 AP4003705 AP4004705 AP4007000 AP4009000 AP4014250 APFB02500 APFC02500 APFD02500 APFA04700 APFA09050 APFB04700 APFC04700 APFC09050 APFD04700 APFD09050 APFF04700 APFF09050 APFF02500 AP4002500 AP4004700 APFB03700 APFB15050 APFC03700 APFF12450 APFF14250 AP408X105
AP4001000EA 52.9 EUR
AP4001000 AP4002405 AP4003705 AP4004705 AP4007000 AP4009000 AP4014250 APFB02500 APFC02500 APFD02500 APFA04700 APFA09050 APFB04700 APFC04700 APFC09050 APFD04700 APFD09050 APFF04700 APFF09050 APFF02500 AP4002500 AP4004700 APFB03700 APFB15050 APFC03700 APFF12450 APFF14250 MILFAP408X105
Glass fiber filters without binder resin
Filters Glass Fibre Filters
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
  • Can be sterilised by EO, gamma, or autoclave.
Type APFA: Retains fine particles with good efficiency, even at high flow rates, used for monitoring wastewater and collecting suspended particles in gases, also have biochemical applications like collection of cells and filtration of protein or nucleic acid precipitates.
Type APFB: Higher mechanicalMore Product Information