Recombinant peroxidase

BIB3098-10KU
APOSBIB3098-10KUEA 77.1 EUR
APOSBIB3098-10KU
Recombinant peroxidase
Enzymes

Order Now

SPECIFICATIONS

Enzyme name Peroxidase
Enzyme synonyms POD, Horseradish peroxidase, Donor: hydrogen-peroxide oxidoreductase
Environmentally Preferable
Enzyme type Recombinant
Source
Species
MDL No. MFCD00071339
Storage conditions Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR