5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98%

Supplier: Apollo Scientific
Warning

BIMB1302-1G BIMB1302-250MG BIMB1302-500MG
APOSBIMB1302-1GEA 232 EUR
APOSBIMB1302-1G APOSBIMB1302-250MG APOSBIMB1302-500MG
5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98%
5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt

Alternative to 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide(X-GlcA) for detection of b-Glucuronidase gene.

Formula: C₁₄H₁₃BrClNO₇
MW: 521.79 g/mol
Melting Pt: 216…217 °C
Storage Temperature: Freezer
MDL Number: MFCD00153929
CAS Number: 144110-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR