Uridine 99%

Supplier: Apollo Scientific
Warning

BIU0400-100G BIU0400-250G BIU0400-25G
APOSBIU0400-100GEA 79.9 EUR
APOSBIU0400-100G APOSBIU0400-250G APOSBIU0400-25G
Uridine 99%
Uridine
Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 163…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006526
CAS Number: 58-96-8
EINECS: 200-407-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR