Aluminium potassium sulphate dodecahydrate ACS

Supplier: Apollo Scientific
Warning

IN1042-2.5KG
APOSIN1042-2.5KGEA 232 EUR
APOSIN1042-2.5KG
Aluminium potassium sulphate dodecahydrate ACS
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92 °C
Density: 1.757 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR