Europium(III) chloride, anhydrous 99,9%

Supplier: Apollo Scientific
Warning

IN1765-25G
APOSIN1765-25GEA 1110 EUR
APOSIN1765-25G
Europium(III) chloride, anhydrous 99,9%
Europium (III) chloride
Formula: Cl₃Eu
MW: 258.32 g/mol
Melting Pt: 850 °C
Density: 4.89 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010994
CAS Number: 10025-76-0
EINECS: 233-040-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR