Manganese(II) chloride tetrahydrate 99%

Supplier: Apollo Scientific
Warning

IN2521-100G
APOSIN2521-100GEA 26.1 EUR
APOSIN2521-100G
Manganese(II) chloride tetrahydrate 99%
Manganese (II) chloride tetrahydrate
Formula: MnCl₂·4H₂O
Boiling Pt: 106 °C
Melting Pt: 58 °C
Density: 2,01 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR