Dipotassium tetraborate tetrahydrate 99%

Supplier: Apollo Scientific
Warning

IN2943-2KG IN2943-500G
APOSIN2943-2KGEA 0 EUR
APOSIN2943-2KG APOSIN2943-500G
Dipotassium tetraborate tetrahydrate 99%
di-Potassium tetraborate tetrahydrate
Formula: B₄K₂O₇·4H₂O
MW: 305.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149924
CAS Number: 12045-78-2
EINECS: 215-575-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR