Tin(II) acetate

IN3634-5G
APOSIN3634-5GEA 80 EUR
APOSIN3634-5G
Tin(II) acetate
Tin (II) acetate
Formula: C₄H₆O₄Sn
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00054408
CAS Number: 638-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR