Potassium-tert-butoxide 97%

Supplier: Apollo Scientific
Danger

OR11033-100G OR11033-500G OR11033-1KG
APOSOR11033-100GEA 102 EUR
APOSOR11033-100G APOSOR11033-500G APOSOR11033-1KG
Potassium-tert-butoxide 97%
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Melting Pt: 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR