2-Amino-3-chloro-5-iodopyrazine

Supplier: Apollo Scientific
Warning

OR300251-1G OR300251-250MG
APOSOR300251-1GEA 144 EUR
APOSOR300251-1G APOSOR300251-250MG
2-Amino-3-chloro-5-iodopyrazine
2-Amino-3-chloro-5-iodopyrazine
Formula: C₄H₃ClIN₃
Storage Temperature: Ambient
MDL Number: MFCD14706048
CAS Number: 1252597-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR