3-Amino-1H-indazole

Supplier: Apollo Scientific
Warning

OR30984-1G OR30984-5G
APOSOR30984-1GEA 21.9 EUR
APOSOR30984-1G APOSOR30984-5G
3-Amino-1H-indazole
3-Amino-1H-indazole
Formula: C₇H₇N₃
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00182045
CAS Number: 874-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR