Triethylene glycol monobenzyl ether 98%

Supplier: Apollo Scientific
Warning

OR3392-25G
APOSOR3392-25GEA 55.1 EUR
APOSOR3392-25G
Triethylene glycol monobenzyl ether 98%
Triethylene glycol monobenzyl ether
Formula: C₁₃H₂₀O₄
MW: 240.30 g/mol
Boiling Pt: 126 °C/0.075mmHg
Density: 1.09 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093556
CAS Number: 55489-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR