4-((3-Hydroxypropyl)amino)-3-nitrophenol 95+%

Supplier: Apollo Scientific

OR350491-100G OR350491-25G
APOSOR350491-100GEA 109 EUR
APOSOR350491-100G APOSOR350491-25G
4-((3-Hydroxypropyl)amino)-3-nitrophenol 95+%
4-((3-Hydroxypropyl)amino)-3-nitrophenol
Formula: C₉H₁₂N₂O₄
MW: 212.2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368637
CAS Number: 92952-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR