Uracil-6-carboxaldehyde monohydrate 98%

Supplier: Apollo Scientific
Warning

OR4145-250MG
APOSOR4145-250MGEA 125 EUR
APOSOR4145-250MG
Uracil-6-carboxaldehyde monohydrate 98%
Uracil-6-carboxaldehyde monohydrate
Formula: C₅H₄N₂O₃·H₂O
MW: 158.112 g/mol (140.10 anhy)
Storage Temperature: Ambient
MDL Number: MFCD03002359
CAS Number: 1052405-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR