4-Bromobenzhydrol 96%

Supplier: Apollo Scientific
Warning

OR4692-1G
APOSOR4692-1GEA 21.9 EUR
APOSOR4692-1G
4-Bromobenzhydrol 96%
4-Bromobenzhydrol
Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00016851
CAS Number: 29334-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR