Methyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate

Supplier: Apollo Scientific

OR470034-5G
APOSOR470034-5GEA 367 EUR
APOSOR470034-5G
Methyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Methyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Formula: C₁₂H₁₂N₂O₃
MW: 232.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198871
CAS Number: 1166756-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR