Cbz-D-Ser-OH 98%

Supplier: Apollo Scientific

OR471601-100G
APOSOR471601-100GEA 75.4 EUR
APOSOR471601-100G
Cbz-D-Ser-OH 98%
Cbz-D-Ser-OH
Formula: C₁₁H₁₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00063144
CAS Number: 6081-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR