3-Chlorophenol

Supplier: Apollo Scientific
Warning

OR8225-100G OR8225-25G OR8225-500G
APOSOR8225-100GEA 52.2 EUR
APOSOR8225-100G APOSOR8225-25G APOSOR8225-500G
3-Chlorophenol
3-Chlorophenol
Formula: C₆H₅ClO
Boiling Pt: 214 °C
Melting Pt: 30…34 °C
Density: 1,245 g/cm³ (25 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002256
CAS Number: 108-43-0
EINECS: 203-582-6
UN: 2020
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR