Sodium butyrate

Supplier: Apollo Scientific
Warning

OR8783-100G OR8783-1KG OR8783-500G
APOSOR8783-100GEA 21.9 EUR
APOSOR8783-100G APOSOR8783-1KG APOSOR8783-500G
Sodium butyrate
Sodium butyrate
Formula: C₄H₇NaO₂
MW: 110.09 g/mol
Melting Pt: 250…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00002816
CAS Number: 156-54-7
EINECS: 205-857-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR