1-(2-Nitrophenyl)ethanol 95%

Supplier: Apollo Scientific

OR953402-5G
APOSOR953402-5GEA 91.5 EUR
APOSOR953402-5G
1-(2-Nitrophenyl)ethanol 95%
1-(2-Nitrophenyl)ethanol
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683380
CAS Number: 3205-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR