Pyrimidine-2-sulphonyl fluoride

Supplier: Apollo Scientific
Danger

PC1172-5G
APOSPC1172-5GEA 909 EUR
APOSPC1172-5G
Pyrimidine-2-sulphonyl fluoride
Pyrimidine-2-sulphonyl fluoride
Formula: C₄H₃FN₂O₂S
MW: 162.14 g/mol
Melting Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD09265503
CAS Number: 35762-87-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR