Barium hexafluorosilicate 98%

Supplier: Apollo Scientific

PC2147-50G
APOSPC2147-50GEA 241 EUR
APOSPC2147-50G
Barium hexafluorosilicate 98%
Barium hexafluorosilicate
Formula: BaF₆Si
Storage Temperature: Ambient
MDL Number: MFCD00014185
CAS Number: 17125-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR