Potassium(morpholin-4-ylmethyl)trifluoroborate

Supplier: Apollo Scientific

PC430508-1G PC430508-5G
APOSPC430508-1GEA 111 EUR
APOSPC430508-1G APOSPC430508-5G
Potassium(morpholin-4-ylmethyl)trifluoroborate
Potassium (morpholin-4-ylmethyl)trifluoroborate
Formula: C₅H₁₀BF₃KNO
MW: 207.0435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566512
CAS Number: 936329-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR