Dimethyl 2-(2-Fluoro-4-nitrophenyl)malonate

Supplier: Apollo Scientific

PC510005-5G
APOSPC510005-5GEA 397 EUR
APOSPC510005-5G
Dimethyl 2-(2-Fluoro-4-nitrophenyl)malonate
Dimethyl 2-(2-Fluoro-4-nitrophenyl)malonate
Formula: C₁₁H₁₀FNO₆
MW: 271.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198898
CAS Number: 188975-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR