Silver hydrogen fluoride 99%

Supplier: Apollo Scientific
Danger

PC6350-5G
APOSPC6350-5GEA 0 EUR
APOSPC6350-5G
Silver hydrogen fluoride 99%
Silver hydrogen fluoride
Formula: AgF₂H
MW: 146.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042145
CAS Number: 12249-52-4
UN: 1740
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR