1-Fluoro-2-isopropoxy-4-nitrobenzene 95%

Supplier: Apollo Scientific

PC902094-1G
APOSPC902094-1GEA 415 EUR
APOSPC902094-1G
1-Fluoro-2-isopropoxy-4-nitrobenzene 95%
1-Fluoro-2-isopropoxy-4-nitrobenzene
Formula: C₉H₁₀FNO₃
MW: 199.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114386
CAS Number: 1229647-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR