Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected 95%

Supplier: Apollo Scientific
Warning

PC909040-1G PC909040-5G
APOSPC909040-1GEA 29 EUR
APOSPC909040-1G APOSPC909040-5G
Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected 95%
Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected
Formula: C₁₀H₁₉BF₃N₂O₂·K
MW: 306.175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566514
CAS Number: 936329-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR