Membrane filters, Isopore™
Supplier: Merck
GTTP04700 TSTP02500 VCTP04700 ATTP01300 ATTP03700 ATTP14250 DTTP01300 DTTP02500 DTTP04700 GTBP01300 GTTP01300 GTTP03700 GTTP09030 GTTP14250 HTBP01300 HTTP03700 HTTP09030 HTTP14250 RTTP01300 RTTP14250 TCTP01300 TCTP02500 TCTP04700 TCTP14250 TETP01300 TETP02500 TETP04700 TETP14250 TMTP01300 TMTP09030 TMTP14250 TSTP01300 TSTP14250 TTTP02500 VCTP01300 VCTP02500 VCTP14250 GTTP02500 GTBP02500 GTBP04700 HTTP02500 HTTP04700 HTBP02500 HTBP04700 ATTP02500 ATTP04700 RTTP02500 RTTP04700 TSTP04700 TMTP02500 TMTP04700 HTTP01300 TTTP04700 VCTP07625 TMTP0190R
GTTP04700EA 444 EUR
GTTP04700 TSTP02500 VCTP04700 ATTP01300 ATTP03700 ATTP14250 DTTP01300 DTTP02500 DTTP04700 GTBP01300 GTTP01300 GTTP03700 GTTP09030 GTTP14250 HTBP01300 HTTP03700 HTTP09030 HTTP14250 RTTP01300 RTTP14250 TCTP01300 TCTP02500 TCTP04700 TCTP14250 TETP01300 TETP02500 TETP04700 TETP14250 TMTP01300 TMTP09030 TMTP14250 TSTP01300 TSTP14250 TTTP02500 VCTP01300 VCTP02500 VCTP14250 GTTP02500 GTBP02500 GTBP04700 HTTP02500 HTTP04700 HTBP02500 HTBP04700 ATTP02500 ATTP04700 RTTP02500 RTTP04700 TSTP04700 TMTP02500 TMTP04700 HTTP01300 TTTP04700 MILFVCTP07625 MILFTMTP0190R
Membrane filters, Isopore™
Filters Membrane Filters
These PC hydrophilic membrane are track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. They are composed of polycarbonate film, which has a smooth, glass like surface for clearer sample observation.
  • Membrane for microscopy and visual analysis
  • Membrane structure retains particles on the surface, simplifying counting and analysis
  • Do not stain, resulting in low
More Product Information