Serotonin hydrochloride ≥98%

Supplier: ALFA AESAR
Danger

B21263.03 B21263.06 B21263.09 B21263.A1
B21263.03EA 126 EUR
B21263.03 B21263.06 B21263.09 B21263.A1
Serotonin hydrochloride ≥98%
Serotonin hydrochloride

Purity: 98% CAS No.: 153-98-0 Molecular Formula: C10H12N2O.HCl Formula Weight: 212.68 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Light Sensitive and Hygroscopic Appearance: White to pale cream Melting Point: 167-172° Beilstein Reference: 3570963 Merck Reference: 14,8463 UN No.: 2811 MDL No.: MFCD00012686

Formula: C₁₀H₁₃ClN₂O
Melting Pt: 167…172 °C
MDL Number: MFCD00012686
CAS Number: 153-98-0
EINECS: 244-464-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,8463

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR