SONOREX DIGITEC DT 514, ultrasonic bath, 230 V( 10%) 50/60 Hz, ID 325x300x150 mm 13,5 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15,30 min and continuous, ultrasonic peak output 860 W, HF output 215 Weff., SweepTec and DEGAS, drain: ball va 1 * 1 items
Supplier: BANDELIN-ELECTRONIC
SONOREX DIGITEC DT 514, ultrasonic bath, 230 V( 10%) 50/60 Hz, ID 325x300x150 mm 13,5 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15,30 min and continuous, ultrasonic peak output 860 W, HF output 215 Weff., SweepTec and DEGAS, drain: ball va 1 * 1 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 88 22222.