Experimentation kits, assays for protein quantification
Supplier: G-Biosciences
BE-402C BE-402B BE-402 BE-402L
BE-402CEA 372 EUR
BE-402C BE-402B BE-402 BE-402L
Experimentation kits, assays for protein quantification
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Determination of protein concentration is an essential technique in all aspects of protein studies and proteomics. The Assays for Protein Quantification kit includes three of the most widely used protein assays and allows for a direct comparison of the three assays that teaches students the benefits and limitations of each assay. Each assay is available individually to allow teaching of a specific assay, without the option of comparing and contrasting with other assays. The three assays covered are the Biuret Protein Assay, Lowry Protein Assay and the Coomassie Blue Dye Protein Assay. TheMore Product Information