Digital polarimeters, ADP600
Supplier: BELLINGHAM STANLEY
37-61-61-181 37-66-61-193 37-61-61-182 37-62-61-181 37-62-61-182 37-62-61-193 37-63-61-181 37-63-61-182 37-63-61-193 37-64-61-181 37-64-61-182 37-64-61-193 37-65-61-181 37-65-61-182 37-65-61-193 37-66-61-181 37-66-61-182 37-61 37-62 37-63 37-64 37-65 37-66
635-0769EA 34560 EUR
635-0769 635-0792 635-0770 635-0773 635-0774 635-0776 635-0777 635-0778 635-0780 635-0781 635-0782 635-0784 635-0785 635-0786 635-0788 635-0789 635-0790 BELL37-61 BELL37-62 BELL37-63 BELL37-64 BELL37-65 BELL37-66
Digital polarimeters, ADP600
Polarimeters
The ADP600 high precision polarimeters with internal Peltier temperature control are capable of measuring optical rotation to four decimal places angular across the UV/Visible spectrum using standard glass or low volume leur taper flow-through tubes.
  • Touch screen facilitates navigation through user and supervisor menus
  • Standard and low volume sample tubes
  • Single, double, or multiple wavelengths for UV/Visible applications
  • GLP software including date/time/batch
Scientists wishing to measure optical rotation, specific rotation or concentration ofMore Product Information