Avantor® ACE® UltraCore BIO phenyl2, Analytical HPLC/UHPLC Columns
Supplier: HICHROM - OEM
BIO-252-521GEA 637 EUR
BIO-252-521G BIO-252-0521 BIO-252-1021 BIO-252-1521 BIO-252-530G BIO-252-0530 BIO-252-1030 BIO-252-1530 BIO-252-546G BIO-252-0546 BIO-252-1046 BIO-252-1546 BIO-352-521G BIO-352-0521 BIO-352-1021 BIO-352-1521 BIO-352-530G BIO-352-0530 BIO-352-1030 BIO-352-1530 BIO-352-546G BIO-352-0546 BIO-352-1046 BIO-352-1546
Avantor® ACE® UltraCore BIO phenyl2, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® UltraCore BIO Phenyl2 are high performing solid-core (core-shell) particle columns, designed for a range of biomolecule applications. With both 300 Å and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of hydrophobic proteins over 5kDa molecular weight with alternative selectivity to C4 bonded phases
  • Wide pore, solid-core combination for excellent peak shape & resolution of large molecules
  • High purity, base deactivated silica with excellent reproducibility
  • Alternative selectivity to
More Product Information