Anti-INS Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB109013-100
BIRBORB109013-100EA 0 EUR
BIRBORB109013-100
Anti-INS Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR