Anti-SYCE1L Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Biorbyt
ORB113770-100
BIRBORB113770-100EA 0 EUR
BIRBORB113770-100
Anti-SYCE1L Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Type: Primary
Antigen: SYCE1L (synaptonemal complex central element protein 1-like)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR