Anti-NTRK2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB131732-100
BIRBORB131732-100EA 0 EUR
BIRBORB131732-100
Anti-NTRK2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: NTRK2 (neurotrophic tyrosine kinase, receptor, type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR