Anti-ELL2 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB131763-100
BIRBORB131763-100EA 0 EUR
BIRBORB131763-100
Anti-ELL2 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: ELL2 (elongation factor, RNA polymerase II, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR