Anti-IQCA1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB1937-100 ORB1937-200
BIRBORB1937-100EA 0 EUR
BIRBORB1937-100 BIRBORB1937-200
Anti-IQCA1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: IQCA1 (IQ motif containing with AAA domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR