Anti-PFKFB2 Goat Polyclonal Antibody

Supplier: Biorbyt
ORB20339-100
BIRBORB20339-100EA 0 EUR
BIRBORB20339-100
Anti-PFKFB2 Goat Polyclonal Antibody
Antibodies
ELISA: 1:16000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: PFKFB2 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR