Anti-Bence Jones Kappa Mouse Monoclonal Antibody (TRITC (Rhodamine))

Supplier: Biorbyt
ORB21181-0.2
BIRBORB21181-0.2EA 0 EUR
BIRBORB21181-0.2
Anti-Bence Jones Kappa Mouse Monoclonal Antibody (TRITC (Rhodamine))
Antibodies
Type: Primary
Antigen: Bence Jones Kappa
Clonality: monoclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR