Anti-NCR1 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB22891-100 ORB22891-20 ORB22891-5
BIRBORB22891-100EA 0 EUR
BIRBORB22891-100 BIRBORB22891-20 BIRBORB22891-5
Anti-NCR1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: NCR1 (natural cytotoxicity triggering receptor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR