Anti-SETDB1 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB69343-100
BIRBORB69343-100EA 0 EUR
BIRBORB69343-100
Anti-SETDB1 Mouse Monoclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; ELISA: 1:10000

Type: Primary
Antigen: SETDB1 (SET domain, bifurcated 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR