Anti-ACOX3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB69795-100
BIRBORB69795-100EA 0 EUR
BIRBORB69795-100
Anti-ACOX3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACOX3 (acyl-CoA oxidase 3, pristanoyl)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ACOX3
Antigen name acyl-CoA oxidase 3, pristanoyl
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
ImmunoChemistry Yes
Concentration 1 mg/ml
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR