Anti-Streptavidin Streptomysces Avidinii Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB79501-1
BIRBORB79501-1EA 0 EUR
BIRBORB79501-1
Anti-Streptavidin Streptomysces Avidinii Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Streptavidin Streptomysces Avidinii
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR