Anti-DRD1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB85693-50
BIRBORB85693-50EA 0 EUR
BIRBORB85693-50
Anti-DRD1 Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 1-5 ?g/ml

Type: Primary
Antigen: DRD1 (dopamine receptor D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR